Learn More
Medchemexpress LLC Ranitidine | 66357-35-5 | MFCD00081180 | 99.9% | 5 MG
SDP
Catalog No. 5000405500
Encompass_Preferred
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It antagonizes histamine-induced increases in guinea-pig isolated rat atrium and histamine-induced relaxations of the rat isolated uterine horn. Studies also show it inhibits breast tumor development and spread in mice.
  • Potent and selective histamine H2-receptor antagonist
  • Orally active
  • Inhibits gastric secretion
  • Antagonizes histamine-induced increases in guinea-pig isolated rat atrium
  • Antagonizes histamine-induced relaxations of rat isolated uterine horn
  • Inhibits breast tumor development and spread in mice
  • Reduces CD11b+Ly6Chi cells in spleen and bone marrow
  • Inhibits lung metastasis
  • Inhibits primary tumor growth
  • Increases latency of breast tumorigenesis
  • Reduces number of tumors
Catalog No. 50-004-05500 Supplier Medchemexpress LLC Supplier No. HYB06935MG
Ranitidine is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It antagonizes histamine-induced increases in guinea-pig isolated rat atrium and histamine-induced relaxations of the rat isolated uterine horn. Studies also show it inhibits breast tumor development and spread in mice.
  • Potent and selective histamine H2-receptor antagonist
  • Orally active
  • Inhibits gastric secretion
  • Antagonizes histamine-induced increases in guinea-pig isolated rat atrium
  • Antagonizes histamine-induced relaxations of rat isolated uterine horn
  • Inhibits breast tumor development and spread in mice
  • Reduces CD11b+Ly6Chi cells in spleen and bone marrow
  • Inhibits lung metastasis
  • Inhibits primary tumor growth
  • Increases latency of breast tumorigenesis
  • Reduces number of tumors