Learn More
Medchemexpress LLC HY-B0281A 100mg Medchemexpress, Ranitidine (hydrochloride) CAS:66357-59-3 Purity:>98%
SDP
Catalog No. 502082445
Encompass_Preferred
Medchemexpress, HY-B0281A 100mg Ranitidine (hydrochloride) CAS:66357-59-3 Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion. Ranitidine hydrochloride is a weak inhibitor of CYP2C19 and CYP2C9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2445 Supplier Medchemexpress LLC Supplier No. HYB0281A100MG
Medchemexpress, HY-B0281A 100mg Ranitidine (hydrochloride) CAS:66357-59-3 Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion. Ranitidine hydrochloride is a weak inhibitor of CYP2C19 and CYP2C9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.