Learn More
Medchemexpress LLC Ranitidine hydrochloride | 66357-59-3 | 99.9% | 500 MG
SDP
Catalog No. 5000406605
Encompass_Preferred
Ranitidine hydrochloride is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It has demonstrated efficacy in antagonizing histamine-induced effects in various in vitro models and has also shown activity in inhibiting breast tumor development and spread in mice.
  • Potent, selective, and orally active histamine H2-receptor antagonist
  • Inhibits gastric secretion
  • Antagonizes histamine-induced increases and relaxations in various in vitro models
  • Inhibits breast tumor development and spread in mice
Catalog No. 50-004-06605 Supplier Medchemexpress LLC Supplier No. HYB0281A500MG
Ranitidine hydrochloride is a potent, selective, and orally active histamine H2-receptor antagonist that inhibits gastric secretion. It has demonstrated efficacy in antagonizing histamine-induced effects in various in vitro models and has also shown activity in inhibiting breast tumor development and spread in mice.
  • Potent, selective, and orally active histamine H2-receptor antagonist
  • Inhibits gastric secretion
  • Antagonizes histamine-induced increases and relaxations in various in vitro models
  • Inhibits breast tumor development and spread in mice