Learn More
Abcam Ranitidine hydrochloride, Selective H2 antagonist, 10MG
Supplier:  Abcam AB12073810MG
Encompass_Preferred
MW 350.86 g/mol, Purity >99%. Potent, competitive and selective H₂ antagonist (pA₂ = 6.95 - 7.2). Inhibits gastric acid secretion in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7318