Learn More
AdipoGen RANK ectodomainh pAb
SDP
Supplier:  AdipoGen AG25A0021C050
Encompass_Preferred
Polyclonal Antibody. Recognizes human RANK. Detects a band of ~24kDa by Western blot. Source Rabbit. Applications FACS, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. RANK is the receptor for RANKL/TRANCE/OPGL. It is essential for RANKL-mediated osteoclastogenesis and is involved in the regulation of interactions between T-cells and dendritic cells.
Catalog No. 50-256-3290