Learn More
Medchemexpress LLC RANK L/TNFSF11 Mous 2ug
SDP
Catalog No. 5000178822
Encompass_Preferred
RANKL (TNFSF11) a type II transmembrane protein is a receptor activator of NF- B (RANK) ligand RANKL is an activator of RANK When binding to RANK it induces the differentiation of monocyte/macrophage-lineage cells into osteoclasts and leads to osteoclast precursor maturation RANKL is critical for osteoclasts maturation bone modeling and bone remodeling as well as the development of lymph nodes (LNs) RANKL/TNFSF11 Protein Mouse is a recombinant mouse RANKL (Q137-D316) without tag which is produced in E coli[1][2]
Catalog No. 50-001-78822 Supplier Medchemexpress LLC Supplier No. HYP74252UG
RANKL (TNFSF11) a type II transmembrane protein is a receptor activator of NF- B (RANK) ligand RANKL is an activator of RANK When binding to RANK it induces the differentiation of monocyte/macrophage-lineage cells into osteoclasts and leads to osteoclast precursor maturation RANKL is critical for osteoclasts maturation bone modeling and bone remodeling as well as the development of lymph nodes (LNs) RANKL/TNFSF11 Protein Mouse is a recombinant mouse RANKL (Q137-D316) without tag which is produced in E coli[1][2]