Learn More
AdipoGen RANK (mouse):Fc (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0092C050
Encompass_Preferred
Protein. Mouse RANK (aa 31-214) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to mouse and human RANKL. Purity: >95% (SDS-PAGE). RANK (TRANCE-R; CD265) is the receptor for RANKL/TRANCE/OPGL. It is essential for RANKL-mediated osteoclastogenesis and is involved in the regulation of interactions between T cells and dendritic cells.
Catalog No. 50-205-3155