Learn More
Sklar Instruments Econo Rankin-Crile Forceps, Sklar, 6-1/4 in., Blunt, Curved, Serrated, Stainless Steel. PK/12.
SDP
Supplier:  Sklar Instruments 21463
Encompass_Preferred
Econo Rankin-Crile Forceps, Sklar, 6-1/4 in., Blunt, Curved, Serrated, Stainless Steel. Econo Rankin-Crile Hemostatic Forceps are a standard clamp regularly used to hold tissue or lumen. Primarily designed to stop bleeding, hemostatic clamps are generally differentiated by weight, size, and serration pattern. Econo Rankin-Crile Hemostatic Clamps feature a jaw that is 1/3 the length of the shank, similar to the Rankin Hemostat, but have serrations the entire length of the jaw, as seen in the Crile Hemostat. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with serrated tips, and an overall length of 6-1/4 inches. They are sold in boxes of 12.
Catalog No. 50-143-7638