Learn More
Stoelting™ Rankin Forceps
SDP
Catalog No. 10001336 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-336 Straight
10-001-337 Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-336 Supplier Stoelting™ Supplier No. 5211018
Only null left
Add to Cart
Add to Cart
Hemostatic forceps are used in surgical procedures to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Specifications
Length (English) 6.25 in.
Model Straight