Learn More
Enzo Life Sciences RANKL (human) monoclonal antibody (Ranky-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804830C100
Encompass_Preferred
Clone: Ranky-1. Isotype: IgG1. Immunogen: Recombinant human soluble RANKL extracellular domain (aa 151-316) (ALX-522-012). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: ELISA, IHC (PS), WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9990