Learn More
Enzo Life Sciences RANKL monoclonal antibody (12A380) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804244C100
Encompass_Preferred
Clone: 12A380. Isotype: IgG1. Immunogen: Recombinant mouse RANKL (aa 1-317). Species reactivity: Human, Mouse. Applications: Flow Cytometry, IHC (FS), WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide.
Catalog No. 50-200-9841