Learn More
Sino Biological RANKL / OPGL / TNFSF11 / CD254 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11682R128100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human RANKL / OPGL / TNFSF11 / CD254 (rh RANKL / OPGL / TNFSF11 / CD254; Catalog#11682-HNCH; AAC51762.1;Gly64-Asp245).
Catalog No. 50-224-7888