Learn More
Sino Biological RANKL / OPGL / TNFSF11 / CD254 Neutralizing Antibody 200ug
Supplier:  Sino Biological 11682R001200
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human RANKL / OPGL / TNFSF11 / CD254 (rh RANKL / OPGL / TNFSF11 / CD254; Catalog#11682-HNCH; AAC51762.1;Gly64-Asp245) and was produced using recombinant antibody technology.
Catalog No. 50-224-7721