Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP540210UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Mouse. BioActivity: Mouse RANKL / OPGL / TNFSF11 / CD254 induces TNF-alpha production in RAW cells (typical ED50 is 50-75 ng/ml). Activity was further confirmed by measuring Osteoclast formation (significanty different from control at Mouse RANKL / OPGL / TNFSF11 / CD254 concentrations greater than 50 ng/ml). AA Sequence: MPAMMEGSWL DVAQRGKPEA QPFAHLTINA ASIPSGSHKV TLSSWYHDRG WAKISNMTLS NGKLRVNQDG FYYLYANICF RHHETSGSVP TDYLQLMVYV VKTSIKIPSS HNLMKGGSTK NWSGNSEFHF YSINVGGFFK LRAGEEISIQ VSNPSLLDPD QDATYFGAFK VQDID. Buffer: 10 mM sodium phosphate and 20 mM sodium chloride, pH 7.5. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: TNFSF11. Gene ID: 21943. Uniprot: O35235.
Catalog No. 50-188-5811