Learn More
AdipoGen RANKL Soluble hrec
SDP
Supplier:  AdipoGen AG40B00083010
Encompass_Preferred
Protein. Human RANKL aa 152-317 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds to human and mouse RANK. Purity >95% SDS-PAGE. RANKL is an osteoclast differentiation and activation factor. Augments the ability of dendritic cells to stimulate naive T cell proliferation. Important regulator of interactions between T cells and dendritic cells and may play a role in the regulation of the T cell-dependent immune response. RANKL plays an important role in the progression of breast cancer.
Catalog No. 50-256-2940