Learn More
Enzo Life Sciences RANKL (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522012C010
Encompass_Preferred
Alternative name: TRANCE, OPGL, ODF, TNFSF 11, CD254. Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human RANKL (aa 152-317) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG-tag. Solubility: Soluble in water. Specificity: Binds to human and mouse RANK. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9362