Learn More
Sino Biological RANKL/TNFSF11 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11682MM43100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RANKL/TNFSF11 (rh RANKL/TNFSF11; Catalog#11682-HNCH; AAC51762.1;Gly64-Asp245). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8643