Learn More
ACROBiosystems Recombinant Protein;Human RANK / TNFRSF11A Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;RAK,CD265,FEO,LOH18CR1,ODFR,OFE,OPTB7,OSTS,PDB2,RANK,TNFRSF11A,TRANCER,CD265 antigen,ODFROSTS,RANKLOH18CR1,NFKB activator,RANK/TNFRSF11A
Supplier:  ACROBiosystems RAKH52511MG
Encompass_Preferred
Recombinant Protein;Human RANK / TNFRSF11A Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human RANK, Mouse IgG2a Fc Tag, low endotoxin (RAK-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 30 - Pro 212 (Accession # Q9Y6Q6-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 47.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TNFSF11, His Tag (Cat. No. RAL-H5240) at 2 μg/mL (100 μL/well) can bind Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-7893