Learn More
Medchemexpress LLC HY-B0280 100mg Medchemexpress, Ranolazine CAS:95635-55-5 Purity:>98%
SDP
Catalog No. 502082444
Encompass_Preferred
Medchemexpress, HY-B0280 100mg Ranolazine CAS:95635-55-5 Ranolazine (CVT 303) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine is also a partial fatty acid oxidation (FAO) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2444 Supplier Medchemexpress LLC Supplier No. HYB0280100MG
Medchemexpress, HY-B0280 100mg Ranolazine CAS:95635-55-5 Ranolazine (CVT 303) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine is also a partial fatty acid oxidation (FAO) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.