Learn More
TARGETMOL CHEMICALS INC RANOLAZINE 200MG
SDP
Catalog No. 502576484
Encompass_Preferred
Also available in 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ranolazine (RS 43285-003) is a calcium uptake inhibitor via the sodium/calcium channel used to treat chronic angina. It affects the sodium-dependent calcium channels during myocardial ischemia in rabbits by altering the intracellular sodium level. purity: 98%
Catalog No. 50-257-6484 Supplier TARGETMOL CHEMICALS INC Supplier No. T6633200MG
Also available in 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ranolazine (RS 43285-003) is a calcium uptake inhibitor via the sodium/calcium channel used to treat chronic angina. It affects the sodium-dependent calcium channels during myocardial ischemia in rabbits by altering the intracellular sodium level. purity: 98%