Learn More
Apexbio Technology LLC Ranolazine 2HCl 95635-56-6 10mg
SDP
Catalog No. 5000093497
Encompass_Preferred
Ranolazine 2HCl (CAS 95635-56-6) is a small molecule frequently utilized in cardiovascular research for its antianginal and antiarrhythmic activities It modulates cardiac metabolism by partially inhibiting fatty acid -oxidation thereby increasing reliance on glucose oxidation This metabolic shift is mediated through activation of pyruvate dehydrogenase in ischemic myocytes enhancing glucose utilization during myocardial ischemia Ranolazine 2HCl s ability to alter substrate preference and improve cellular energetics under hypoxic conditions supports its application in studies of cardiac ischemia arrhythmias and myocardial metabolism
Catalog No. 50-000-93497 Supplier Apexbio Technology LLC Supplier No. A530010
Ranolazine 2HCl (CAS 95635-56-6) is a small molecule frequently utilized in cardiovascular research for its antianginal and antiarrhythmic activities It modulates cardiac metabolism by partially inhibiting fatty acid -oxidation thereby increasing reliance on glucose oxidation This metabolic shift is mediated through activation of pyruvate dehydrogenase in ischemic myocytes enhancing glucose utilization during myocardial ischemia Ranolazine 2HCl s ability to alter substrate preference and improve cellular energetics under hypoxic conditions supports its application in studies of cardiac ischemia arrhythmias and myocardial metabolism