Learn More
Apexbio Technology LLC Ranolazine 95635-55-5 200mg
SDP
Catalog No. 5000095157
Encompass_Preferred
Ranolazine (CAS 95635-55-5) is a small molecule that acts as an anti-ischemic agent by selectively inhibiting the late sodium current (INaL) in cardiac cells thereby reducing intracellular sodium accumulation and subsequent calcium overload via the Na /Ca2 exchanger This action mitigates adverse effects associated with ischemia-induced ionic imbalance Ranolazine has also been shown to shift ATP production from fatty acid oxidation toward glucose oxidation reflecting greater metabolic efficiency as glucose oxidation yields more ATP per mole of oxygen consumed Additionally it decreases oxygen consumption and suppresses fatty acid-driven ketogenesis in hepatic cells Ranolazine is widely utilized in research focused on myocardial energy metabolism and ischemic heart disease
Catalog No. 50-000-95157 Supplier Apexbio Technology LLC Supplier No. A8510200
Ranolazine (CAS 95635-55-5) is a small molecule that acts as an anti-ischemic agent by selectively inhibiting the late sodium current (INaL) in cardiac cells thereby reducing intracellular sodium accumulation and subsequent calcium overload via the Na /Ca2 exchanger This action mitigates adverse effects associated with ischemia-induced ionic imbalance Ranolazine has also been shown to shift ATP production from fatty acid oxidation toward glucose oxidation reflecting greater metabolic efficiency as glucose oxidation yields more ATP per mole of oxygen consumed Additionally it decreases oxygen consumption and suppresses fatty acid-driven ketogenesis in hepatic cells Ranolazine is widely utilized in research focused on myocardial energy metabolism and ischemic heart disease