Learn More
Medchemexpress LLC Ranolazine-d3 | 1054624-77-9 | 99.5% | 100 MG
SDP
Catalog No. 5000407053
Encompass_Preferred
Ranolazine-d3 is a deuterium labeled anti-angina agent that inhibits the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure. It also functions as a partial fatty acid oxidation inhibitor and is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07053 Supplier Medchemexpress LLC Supplier No. HYB0280S2100MG
Ranolazine-d3 is a deuterium labeled anti-angina agent that inhibits the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure. It also functions as a partial fatty acid oxidation inhibitor and is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.