Learn More
Medchemexpress LLC Ranolazine-d3 | 1054624-77-9 | 99.5% | 25 MG
SDP
Catalog No. 5000450628
Encompass_Preferred
Ranolazine-d3 is the deuterium labeled version of Ranolazine, an anti-angina agent that inhibits the late phase of inward sodium current (INa) and IKr, with IC50 values of 6 μM and 12 μM, respectively, without impacting heart rate or blood pressure. It also acts as a partial fatty acid oxidation (FAO) inhibitor. This compound is an antianginal agent primarily used for research purposes. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50628 Supplier Medchemexpress LLC Supplier No. 25MGHYB0280S2
Ranolazine-d3 is the deuterium labeled version of Ranolazine, an anti-angina agent that inhibits the late phase of inward sodium current (INa) and IKr, with IC50 values of 6 μM and 12 μM, respectively, without impacting heart rate or blood pressure. It also acts as a partial fatty acid oxidation (FAO) inhibitor. This compound is an antianginal agent primarily used for research purposes. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS