Learn More
Medchemexpress LLC Ranolazine-d3 | 1054624-77-9 | 99.5% | 50 MG
SDP
Catalog No. 5000407054
Encompass_Preferred
Ranolazine-d3 is the deuterium labeled Ranolazine, which is known as an anti-angina agent (CVT 303). It functions by inhibiting the late phase of inward sodium current (INa and IKr) at IC50 values of 6 μM and 12 μM, respectively, without influencing heart rate or blood pressure. Additionally, it serves as a partial fatty acid oxidation (FAO) inhibitor and an antianginal agent.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07054 Supplier Medchemexpress LLC Supplier No. HYB0280S250MG
Ranolazine-d3 is the deuterium labeled Ranolazine, which is known as an anti-angina agent (CVT 303). It functions by inhibiting the late phase of inward sodium current (INa and IKr) at IC50 values of 6 μM and 12 μM, respectively, without influencing heart rate or blood pressure. Additionally, it serves as a partial fatty acid oxidation (FAO) inhibitor and an antianginal agent.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.