Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ranolazine-d3 | 1054624-77-9 | MFCD00864690 | 99.5% | 430.56 | C24H30D3N3O4 | 10 MG
SDP

Catalog No. 5000298283
Encompass_Preferred

Deuterium-labeled ranolazine (d3) for research use as an analytical tracer and internal standard. The compound inhibits the late inward sodium current (INa) and the rapid delayed rectifier potassium current (IKr), and acts as a partial fatty acid oxidation inhibitor; it is intended for method development and quantitative analysis by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled tracer suitable for quantitation and metabolic studies.
  • Purity 99.5%.
  • Molecular formula C24H30D3N3O4; molecular weight 430.56.
  • Soluble ≥100 mg/mL in DMSO for in vitro applications.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-98283 Supplier Medchemexpress LLC Supplier No. HYB0280S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled ranolazine (d3) for research use as an analytical tracer and internal standard. The compound inhibits the late inward sodium current (INa) and the rapid delayed rectifier potassium current (IKr), and acts as a partial fatty acid oxidation inhibitor; it is intended for method development and quantitative analysis by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled tracer suitable for quantitation and metabolic studies.
  • Purity 99.5%.
  • Molecular formula C24H30D3N3O4; molecular weight 430.56.
  • Soluble ≥100 mg/mL in DMSO for in vitro applications.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.