Learn More
Medchemexpress LLC Ranolazine-d8 | 1092804-88-0 | 99.8% | 1 MG
SDP
Catalog No. 5000408548
Encompass_Preferred
Ranolazine-d8 is the deuterium labeled Ranolazine. Ranolazine is an anti-angina agent that inhibits the late phase of inward sodium current without affecting heart rate or blood pressure. It is also a partial fatty acid oxidation inhibitor.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08548 Supplier Medchemexpress LLC Supplier No. HYB0280S11MG
Ranolazine-d8 is the deuterium labeled Ranolazine. Ranolazine is an anti-angina agent that inhibits the late phase of inward sodium current without affecting heart rate or blood pressure. It is also a partial fatty acid oxidation inhibitor.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.