Learn More
Medchemexpress LLC Ranolazine-d8 | 1092804-88-0 | 99.8% | 5 MG
SDP
Catalog No. 5000409094
Encompass_Preferred
Ranolazine-d8 is the deuterium labeled Ranolazine, an anti-angina agent that inhibits the late phase of inward sodium current without affecting heart rate or blood pressure. It is also a partial fatty acid oxidation (FAO) inhibitor. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Inhibits the late phase of inward sodium current
  • Does not affect heart rate or blood pressure
  • Acts as a partial fatty acid oxidation (FAO) inhibitor
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09094 Supplier Medchemexpress LLC Supplier No. HYB0280S15MG
Ranolazine-d8 is the deuterium labeled Ranolazine, an anti-angina agent that inhibits the late phase of inward sodium current without affecting heart rate or blood pressure. It is also a partial fatty acid oxidation (FAO) inhibitor. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Inhibits the late phase of inward sodium current
  • Does not affect heart rate or blood pressure
  • Acts as a partial fatty acid oxidation (FAO) inhibitor
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS