Learn More
Medchemexpress LLC HY-17401 200mg Medchemexpress, Ranolazine (dihydrochloride) CAS:95635-56-6 Purity:>98%
SDP
Catalog No. 502081945
Encompass_Preferred
Medchemexpress, HY-17401 200mg Ranolazine (dihydrochloride) CAS:95635-56-6 Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1945 Supplier Medchemexpress LLC Supplier No. HY17401200MG
Medchemexpress, HY-17401 200mg Ranolazine (dihydrochloride) CAS:95635-56-6 Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.