Learn More
TARGETMOL CHEMICALS INC Ranolazine dihydrochloride
SDP
Catalog No. 502880647
Encompass_Preferred
Also available in 1 g, 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ranolazine dihydrochloride (Ranolazine 2HCl) , an antianginal agent, can treat arrhythmia via a novel mechanism of action (inhibition of the late phase of the inward sodium current), and do not affect blood pressure or heart rate. Purity 100%
Catalog No. 50-288-0647 Supplier TARGETMOL CHEMICALS INC Supplier No. T0207500MG
Also available in 1 g, 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Ranolazine dihydrochloride (Ranolazine 2HCl) , an antianginal agent, can treat arrhythmia via a novel mechanism of action (inhibition of the late phase of the inward sodium current), and do not affect blood pressure or heart rate. Purity 100%