Learn More
Medchemexpress LLC Ranolazine (dihydrochloride) | 95635-56-6 | 500.46 | 1 ML
SDP
Catalog No. 5000384268
Encompass_Preferred
Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina agent that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). It is also a partial fatty acid oxidation inhibitor.
  • Functions as an anti-angina agent.
  • Inhibits the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively).
  • Does not affect heart rate or blood pressure (BP).
  • Serves as a partial fatty acid oxidation inhibitor.
Catalog No. 50-003-84268 Supplier Medchemexpress LLC Supplier No. HY1740110MM/1ML
Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina agent that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). It is also a partial fatty acid oxidation inhibitor.
  • Functions as an anti-angina agent.
  • Inhibits the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively).
  • Does not affect heart rate or blood pressure (BP).
  • Serves as a partial fatty acid oxidation inhibitor.