Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ranolazine dihydrochloride | 95635-56-6 | 99.7% | 500.46 | 5 G
SDP

Supplier:  Medchemexpress LLC HY174015G

Encompass_Preferred

Ranolazine dihydrochloride is an anti-angina agent that inhibits the late phase of inward sodium current (INa and IKr) without affecting heart rate or blood pressure. It also acts as a partial fatty acid oxidation inhibitor.

  • Appears as a white to off-white solid
  • High purity
  • Soluble in DMSO, H2O, and PBS
  • Recommended storage at 4°C, sealed and away from moisture
  • Inhibits late phase of inward sodium current and IKr
  • Functions as a partial fatty acid oxidation inhibitor

Catalog No. 50-003-85960


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.