Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ranolazine (dihydrochloride) | 95635-56-6 | 99.99% | 500.46 | 1 G
SDP

Supplier:  Medchemexpress LLC HY174011G

Encompass_Preferred

Ranolazine dihydrochloride is an anti-angina agent that works by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure. It is also a partial fatty acid oxidation inhibitor. It is intended for research use only and is not sold to patients.

  • Anti-angina agent
  • Inhibits late phase of inward sodium current (INa and IKr)
  • Does not affect heart rate or blood pressure
  • Partial fatty acid oxidation inhibitor
  • Purity: 99.99%
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO (≥ 50 mg/mL)
  • Soluble in H2O (≥ 50 mg/mL)

Catalog No. 50-003-85294


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.