Learn More
Abcam Ranolazine dihydrochloride, Anti-ischemic agent, 100MG
Catalog No. 502706149
Encompass_Preferred
MW 500.5 Da, Purity >99%. Novel metabolic modulator and membrane stabilizer. Partial fatty acid oxidation inhibitor. Pyruvate dehydrogenase activator. Shifts myocardial energy metabolism towards glucose for ATP production. Inhibits late INa and IKr currents. Anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6149 Supplier Abcam Supplier No. AB142942100MG
MW 500.5 Da, Purity >99%. Novel metabolic modulator and membrane stabilizer. Partial fatty acid oxidation inhibitor. Pyruvate dehydrogenase activator. Shifts myocardial energy metabolism towards glucose for ATP production. Inhibits late INa and IKr currents. Anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement