Learn More
Selleck Chemical LLC Ranolazine 200mg 95635-55-5 CVT 303, RS 43285-003
SDP
Supplier:  Selleck Chemical LLC S1799200MG
Encompass_Preferred
Ranolazine (CVT 303, RS 43285-003) is a calcium uptake inhibitor via the sodium/calcium channel, used to treat chronic angina. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-661-4