Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RANTES (CCL5) protein, rat | 50 UG
SDP

Catalog No. 5000293374
Encompass_Preferred

Recombinant rat RANTES/CCL5 (residues S25-S92), His-tagged and expressed in Pichia pastoris, supplied as a sterile, 0.22 μm filtered PBS solution (pH 7.4) for use in chemotaxis, receptor binding, and inflammation research. Endotoxin level is <1 EU/μg; store at -80 °C and avoid repeated freeze-thaw cycles.

  • His-tagged recombinant rat chemokine (residues S25-S92).
  • Expression system: Pichia pastoris.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Endotoxin level <1 EU/μg.
  • Pack size: 50 μg vial.
  • Store at -80 °C; aliquot to avoid freeze-thaw.

Catalog No. 50-002-93374 Supplier Medchemexpress LLC Supplier No. HYP7460250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant rat RANTES/CCL5 (residues S25-S92), His-tagged and expressed in Pichia pastoris, supplied as a sterile, 0.22 μm filtered PBS solution (pH 7.4) for use in chemotaxis, receptor binding, and inflammation research. Endotoxin level is <1 EU/μg; store at -80 °C and avoid repeated freeze-thaw cycles.

  • His-tagged recombinant rat chemokine (residues S25-S92).
  • Expression system: Pichia pastoris.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Endotoxin level <1 EU/μg.
  • Pack size: 50 μg vial.
  • Store at -80 °C; aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.