Learn More
Enzo Life Sciences RAP (human), (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5520100
Encompass_Preferred
Alternative name: LDL receptor associated protein. LRPAP1. LDL receptor related protein associated protein 1. MW: ~39kDa (SDS-PAGE). Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 0.0067M Phosphate, pH 7.4 containing 0.15M sodium chloride. Source: Produced in E. coli. UniProt: P30533. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC0073096