Learn More
Enzo Life Sciences Rap1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPGP125E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Rap1a. The sequence is completely conserved in mouse, rat, bovine, chicken, Xenopus. Differs from a portion of Rap1b by 2 amino acids. UniProt ID: P62834 (Rap1A), P61224 (Rap1B). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7276