Learn More
Aviva Systems Biology Corp RAP1GAP Antibody - middle region (ARP56511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAP1GAP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAP1GAP. Alternate names: KIAA0474, RAP1GA1, rap1GAPII, RAPGAP, RAP1GAP1, RAP1GAPIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61887