Learn More
Aviva Systems Biology Corp RAP1GDS1 Antibody - middle region (ARP59654_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59654_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAP1GDS1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAP1GDS1. Alternate names: GDS1, MGC118859, MGC118861, SmgGDSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-769