Learn More
Aviva Systems Biology Corp RAP2A Antibody - N-terminal region (ARP64033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAP2A. It was validated on Western Blot. . Alternate names: K-REV, KREV, RAP2, RbBP-30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-149