Learn More
Aviva Systems Biology Corp RAP2B Antibody - C-terminal region (ARP64354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAP2B. It was validated on Western Blot. . Alternate names: MGC20484ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-243