Learn More
Medchemexpress LLC Rapacuronium bromide | 156137-99-4 | 98.0% | 677.80 | 50 MG
SDP
Catalog No. 5000394473
Encompass_Preferred
Rapacuronium bromide (Org 9487) is a non-depolarizing neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptor (mAChR).
  • Acts as a non-depolarizing neuromuscular blocker
  • Functions as an allosteric modulator of muscarinic acetylcholine receptor (mAChR)
  • Binds to all muscarinic receptor subtypes
  • Exhibits micromolar affinity and slight selectivity towards M2 receptor
  • Lowers [35S]GTPγS binding to membranes
Catalog No. 50-003-94473 Supplier Medchemexpress LLC Supplier No. HY1642350MG
Rapacuronium bromide (Org 9487) is a non-depolarizing neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptor (mAChR).
  • Acts as a non-depolarizing neuromuscular blocker
  • Functions as an allosteric modulator of muscarinic acetylcholine receptor (mAChR)
  • Binds to all muscarinic receptor subtypes
  • Exhibits micromolar affinity and slight selectivity towards M2 receptor
  • Lowers [35S]GTPγS binding to membranes