Learn More
Medchemexpress LLC Rapacuronium bromide | 156137-99-4 | MFCD00870871 | 98.0% | 677.80 g·mol⁻¹ | C37H61BrN2O4 | 10 MG
SDP
Catalog No. 5000289109
Encompass_Preferred
Rapacuronium bromide is a non-depolarizing aminosteroid neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptors supplied for research use only. It is offered in solid and solution formats across multiple pack sizes and is provided at high purity.
  • Non-depolarizing neuromuscular blocker and allosteric modulator of muscarinic acetylcholine receptors.
  • Intended for research use only; not for human or veterinary use.
  • Available in solid and DMSO solution formats for flexible experimental use.
  • High purity (98.0%) suitable for analytical and preclinical studies.
  • Provided with CAS number and chemical formula for traceability.
Catalog No. 50-002-89109 Supplier Medchemexpress LLC Supplier No. HY1642310MG
Rapacuronium bromide is a non-depolarizing aminosteroid neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptors supplied for research use only. It is offered in solid and solution formats across multiple pack sizes and is provided at high purity.
  • Non-depolarizing neuromuscular blocker and allosteric modulator of muscarinic acetylcholine receptors.
  • Intended for research use only; not for human or veterinary use.
  • Available in solid and DMSO solution formats for flexible experimental use.
  • High purity (98.0%) suitable for analytical and preclinical studies.
  • Provided with CAS number and chemical formula for traceability.