Learn More
Medchemexpress LLC Rapacuronium bromide (Org 9487) | 156137-99-4 | 98.0% | 677.80 | 100 MG
SDP
Catalog No. 5000394143
Encompass_Preferred
Rapacuronium bromide (Org 9487) is a non-depolarizing neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptor (mAChR). It is for research use only.
  • Non-depolarizing neuromuscular blocker
  • Allosteric modulator of muscarinic acetylcholine receptor (mAChR)
  • Binds to all muscarinic receptor subtypes with micromolar affinity and slight selectivity towards M2 receptor
  • Exhibits complex effects on the kinetics of ACh binding and subsequent receptor activation
Catalog No. 50-003-94143 Supplier Medchemexpress LLC Supplier No. HY16423100MG
Rapacuronium bromide (Org 9487) is a non-depolarizing neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptor (mAChR). It is for research use only.
  • Non-depolarizing neuromuscular blocker
  • Allosteric modulator of muscarinic acetylcholine receptor (mAChR)
  • Binds to all muscarinic receptor subtypes with micromolar affinity and slight selectivity towards M2 receptor
  • Exhibits complex effects on the kinetics of ACh binding and subsequent receptor activation