Learn More
Medchemexpress LLC Rapacuronium bromide (Org 9487) | 156137-99-4 | 98.0% | 677.80 | 25 MG
SDP
Catalog No. 5000395178
Encompass_Preferred
Rapacuronium bromide (Org 9487) is a non-depolarizing neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptor (mAChR). It is for research use only and not sold to patients.
  • Non-depolarizing neuromuscular blocker
  • Allosteric modulator of muscarinic acetylcholine receptor (mAChR)
Catalog No. 50-003-95178 Supplier Medchemexpress LLC Supplier No. 25MGHY16423
Rapacuronium bromide (Org 9487) is a non-depolarizing neuromuscular blocker and an allosteric modulator of muscarinic acetylcholine receptor (mAChR). It is for research use only and not sold to patients.
  • Non-depolarizing neuromuscular blocker
  • Allosteric modulator of muscarinic acetylcholine receptor (mAChR)