Learn More
Medchemexpress LLC HY-111373 1mg Medchemexpress, RapaLink-1 CAS:1887095-82-0 Purity:>98%
SDP
Catalog No. 502029692
Encompass_Preferred
Medchemexpress, HY-111373 1mg RapaLink-1 CAS:1887095-82-0 RapaLink-1, the third-generation bivalent mTOR inhibitor, combines Rapamycin with MLN0128 (a second-generation mTOR kinase inhibitor) by an inert chemical linker. RapaLink-1 shows better efficacy than Rapamycin or mTOR kinase inhibitors (TORKi), potently blocking cancer-derived, activating mutants of mTOR. RapaLink-1 can cross the blood-brain barrier. RapaLink-1 binding to FKBP12 results in targeted and durable inhibition of mTORC1. RapaLink-1 plays an antithrombotic role in antiphospholipid syndrome by improving autophagy. Anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9692 Supplier Medchemexpress LLC Supplier No. HY1113731MG
Medchemexpress, HY-111373 1mg RapaLink-1 CAS:1887095-82-0 RapaLink-1, the third-generation bivalent mTOR inhibitor, combines Rapamycin with MLN0128 (a second-generation mTOR kinase inhibitor) by an inert chemical linker. RapaLink-1 shows better efficacy than Rapamycin or mTOR kinase inhibitors (TORKi), potently blocking cancer-derived, activating mutants of mTOR. RapaLink-1 can cross the blood-brain barrier. RapaLink-1 binding to FKBP12 results in targeted and durable inhibition of mTORC1. RapaLink-1 plays an antithrombotic role in antiphospholipid syndrome by improving autophagy. Anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.