Learn More
Apexbio Technology LLC Rapalink-1, 5mg Cas No: NA MFCD: NA
SDP
Supplier:  Apexbio Technology LLC A87645
Encompass_Preferred
RapaLink-1 is the third-generation mTOR inhibitor exploiting the unique juxtaposition of two drug (first- and second-generation mTOR kinase inhibitors) binding pockets to create a bivalent interaction that allows inhibition of the mutants which has resistance to the previous TORKi. There are other sizes available. Please inqury us for quote.
Catalog No. NC1535447