Learn More
Gold Biotechnology Inc Rapamycin 100 mg
SDP
Catalog No. 5000562155
Encompass_Preferred
Rapamycin is a macrolide antibiotic isolated from Streptomyces hygroscopicus It has been shown to display anticancer activity such as induction of apoptosis and inhibition of angiogenesis and proliferation in tumor cell lines Rapamycin is also an immunosuppressant and FRAP inhibitor and inhibits lymphokine-induced cell proliferation
Catalog No. 50-005-62155 Supplier Gold Biotechnology Inc Supplier No. R101100
Rapamycin is a macrolide antibiotic isolated from Streptomyces hygroscopicus It has been shown to display anticancer activity such as induction of apoptosis and inhibition of angiogenesis and proliferation in tumor cell lines Rapamycin is also an immunosuppressant and FRAP inhibitor and inhibits lymphokine-induced cell proliferation