Learn More
Cayman Chemical Rapamycin, 53123-88-9, 10 mg
SDP
Supplier:  Cayman Chemical 1334610
Encompass_Preferred
Molecular Formula C51H79NO13, Purity ≥95%, Formula Weight 914.2, Synonyms: Sirolimus; An immunosuppressant used to prevent transplant rejection; potent inhibitor of IL-2 activation of lymphocytes (IC50 = 5 pM); specifically interacts with FKBP12 to form a complex which inhibits mTORC1.
Catalog No. NC0562909